ContactUsLinkComponent
Rotavapor R-300 no bath, SJ29/32, S, I-300P, VU, 230V. . Lift System: Elec. Heating Bath: w/o SJ 29/32 Glass set: Vertical reflux ( S) int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300201S051
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ29/32, S, I-300P, VU, 230V. . Lift System: Elec. Heating Bath: w/o SJ 29/32 Glass set: Vertical reflux ( S) int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300201S051
Supplier:  BUCHI
Frequently Bought Together