ContactUsLinkComponent
Rotavapor R-300 B-305, SJ24/40, V, I-300P, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: Vert ical (V) int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300152V031
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ24/40, V, I-300P, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: Vert ical (V) int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300152V031
Supplier:  BUCHI
Frequently Bought Together