ContactUsLinkComponent
Rotavapor R-300 B-301, SJ29/32, A, I-300, 230V. . Elec. lift, B-301 bath, SJ 29/32 Glass set: Diagonal (A) int.: I-300, Woulff bott le 1 * 1 items
Catalog # BUCH11R300211A021
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ29/32, A, I-300, 230V. . Elec. lift, B-301 bath, SJ 29/32 Glass set: Diagonal (A) int.: I-300, Woulff bott le 1 * 1 items
Catalog # BUCH11R300211A021
Supplier:  BUCHI
Frequently Bought Together