ContactUsLinkComponent
Rotavapor R-300 B-301, SJ29/32, V, P+G, I-300, 230V. . Elec. lift, B-301 bath, SJ 29/32 Glass set: Vertical (V) P+G coating, int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300211V121
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ29/32, V, P+G, I-300, 230V. . Elec. lift, B-301 bath, SJ 29/32 Glass set: Vertical (V) P+G coating, int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300211V121
Supplier:  BUCHI
Frequently Bought Together