ContactUsLinkComponent
Rotavapor R-300 B-301, SJ24/40, A, I-300, 230V. . Elec. lift, B-301 bath, SJ 24/40 Glass set: Diagonal (A) int.: I-300, Woulff bott le 1 * 1 items
Catalog # BUCH11R300212A021
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ24/40, A, I-300, 230V. . Elec. lift, B-301 bath, SJ 24/40 Glass set: Diagonal (A) int.: I-300, Woulff bott le 1 * 1 items
Catalog # BUCH11R300212A021
Supplier:  BUCHI
Frequently Bought Together