ContactUsLinkComponent
Rotavapor R-300 B-305, SJ24/40, V, I-300, VU, 230V. . Elec. lift, B-305 bath, SJ 24/40 Glass set: Vertical (V) int.: I-300, valve u nit 1 * 1 items
Catalog # BUCH11R300252V041
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ24/40, V, I-300, VU, 230V. . Elec. lift, B-305 bath, SJ 24/40 Glass set: Vertical (V) int.: I-300, valve u nit 1 * 1 items
Catalog # BUCH11R300252V041
Supplier:  BUCHI
Frequently Bought Together