ContactUsLinkComponent
Rotavapor R-300 B-301, no joint., V, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) Glass set: Vertical ( V) int.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300110V041
Supplier:  BUCHI
Rotavapor R-300 B-301, no joint., V, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) Glass set: Vertical ( V) int.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300110V041
Supplier:  BUCHI
Frequently Bought Together