ContactUsLinkComponent
Rotavapor R-300 B-301, SJ29/32, S, I-300, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 29/32 Glass set: Vertic al reflux (S) int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300111S021
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ29/32, S, I-300, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 29/32 Glass set: Vertic al reflux (S) int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300111S021
Supplier:  BUCHI
Frequently Bought Together