ContactUsLinkComponent
Rotavapor R-300 B-301, SJ24/40, A, P+G, I-300, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 24/40 Glass set: D iagonal (A) P+G coating, int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300112A121
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ24/40, A, P+G, I-300, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) SJ 24/40 Glass set: D iagonal (A) P+G coating, int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300112A121
Supplier:  BUCHI
Frequently Bought Together