ContactUsLinkComponent
Rotavapor R-300 B-305, no joint., S, I-300, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) Glass set: Vertical refl ux (S) int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300150S021
Supplier:  BUCHI
Rotavapor R-300 B-305, no joint., S, I-300, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) Glass set: Vertical refl ux (S) int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300150S021
Supplier:  BUCHI
Frequently Bought Together