ContactUsLinkComponent
Rotavapor R-300 B-305, no joint., no glass, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) Glass set: w/ o int.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300150N041
Supplier:  BUCHI
Rotavapor R-300 B-305, no joint., no glass, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) Glass set: w/ o int.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300150N041
Supplier:  BUCHI
Frequently Bought Together