ContactUsLinkComponent
Rotavapor R-300 B-305, SJ29/32, A, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 29/32 Glass set: D iagonal (A) int.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300151A041
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ29/32, A, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 29/32 Glass set: D iagonal (A) int.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300151A041
Supplier:  BUCHI
Frequently Bought Together