ContactUsLinkComponent
Rotavapor R-300 B-301, no joint., C, P+G, I-300P, VU, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) Glass set: Cold trap, P+G coating, int.: I-300 Pro, valve unit 1 * 1 items
  BUCH11R300110C151
 :  BUCHI
Rotavapor R-300 B-301, no joint., C, P+G, I-300P, VU, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) Glass set: Cold trap, P+G coating, int.: I-300 Pro, valve unit 1 * 1 items
  BUCH11R300110C151
 :  BUCHI
Frequently Bought Together