ContactUsLinkComponent
Rotavapor R-300 B-305, SJ24/40, C, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: C old trap, int.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300152C041
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ24/40, C, I-300, VU, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: C old trap, int.: I-300, valve unit 1 * 1 items
Catalog # BUCH11R300152C041
Supplier:  BUCHI
Frequently Bought Together