ContactUsLinkComponent
Rotavapor R-300 B-305, SJ24/40, A, I-300, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: Diago nal (A) int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300152A021
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ24/40, A, I-300, 230V. . Lift System: Manual Heating Bath: B-305 (20 - 220?, 5L) SJ 24/40 Glass set: Diago nal (A) int.: I-300, Woulff bottle 1 * 1 items
Catalog # BUCH11R300152A021
Supplier:  BUCHI
Frequently Bought Together