ContactUsLinkComponent
Rotavapor R-300 B-301, no joint., A, P+G, I-300P, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) Glass set: Diagonal (A) P+G coating, int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300110A131
Supplier:  BUCHI
Rotavapor R-300 B-301, no joint., A, P+G, I-300P, 230V. . Lift System: Manual Heating Bath: B-301 (20 - 95?, 1L) Glass set: Diagonal (A) P+G coating, int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300110A131
Supplier:  BUCHI
Frequently Bought Together