ContactUsLinkComponent
Rotavapor R-300 no bath, SJ24/40, S, P+G, I-300P, 230V. . Lift System: Manual Heating Bath: w/o SJ 24/40 Glass set: Vertical reflux (S) P+G coating, int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300102S131
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ24/40, S, P+G, I-300P, 230V. . Lift System: Manual Heating Bath: w/o SJ 24/40 Glass set: Vertical reflux (S) P+G coating, int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300102S131
Supplier:  BUCHI
Frequently Bought Together