ContactUsLinkComponent
Rotavapor R-300 no bath, SJ24/40, C, I-300P, VU, 230V. . Lift System: Manual Heating Bath: w/o SJ 24/40 Glass set: Cold trap, int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300102C051
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ24/40, C, I-300P, VU, 230V. . Lift System: Manual Heating Bath: w/o SJ 24/40 Glass set: Cold trap, int.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300102C051
Supplier:  BUCHI
Frequently Bought Together