ContactUsLinkComponent
Rotavapor R-300 no bath, SJ29/32, no glass, I-300, 230V. . Lift System: Manual Heating Bath: w/o SJ 29/32 Glass set: w/o int.: I-30 0, Woulff bottle 1 * 1 items
Catalog # BUCH11R300101N021
Supplier:  BUCHI
Rotavapor R-300 no bath, SJ29/32, no glass, I-300, 230V. . Lift System: Manual Heating Bath: w/o SJ 29/32 Glass set: w/o int.: I-30 0, Woulff bottle 1 * 1 items
Catalog # BUCH11R300101N021
Supplier:  BUCHI
Frequently Bought Together