ContactUsLinkComponent
Rotavapor R-300 B-305, SJ29/32, V, P+G, I-300P, VU, 230V. . Elec. lift, B-305 bath, SJ 29/32 Glass set: Vertical (V) P+G coating, i nt.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300251V151
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ29/32, V, P+G, I-300P, VU, 230V. . Elec. lift, B-305 bath, SJ 29/32 Glass set: Vertical (V) P+G coating, i nt.: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300251V151
Supplier:  BUCHI
Frequently Bought Together