ContactUsLinkComponent
Rotavapor R-300 B-305, SJ24/40, A, I-300P, VU, 230V. . Elec. lift, B-305 bath, SJ 24/40 Glass set: Diagonal (A) int.: I-300 Pro, va lve unit 1 * 1 items
Catalog # BUCH11R300252A051
Supplier:  BUCHI
Rotavapor R-300 B-305, SJ24/40, A, I-300P, VU, 230V. . Elec. lift, B-305 bath, SJ 24/40 Glass set: Diagonal (A) int.: I-300 Pro, va lve unit 1 * 1 items
Catalog # BUCH11R300252A051
Supplier:  BUCHI
Frequently Bought Together