ContactUsLinkComponent
Rotavapor R-300 B-301, SJ24/40, V, I-300P, 230V. . Elec. lift, B-301 bath, SJ 24/40 Glass set: Vertical (V) int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300212V031
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ24/40, V, I-300P, 230V. . Elec. lift, B-301 bath, SJ 24/40 Glass set: Vertical (V) int.: I-300 Pro, Woulff bottle 1 * 1 items
Catalog # BUCH11R300212V031
Supplier:  BUCHI
Frequently Bought Together