ContactUsLinkComponent
Rotavapor R-300 B-301, SJ24/40, no glass, I-300P, VU, 230V. . Elec. lift, B-301 bath, SJ 24/40 Glass set: w/o int.: I-300 Pro, valv e unit 1 * 1 items
Catalog # BUCH11R300212N051
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ24/40, no glass, I-300P, VU, 230V. . Elec. lift, B-301 bath, SJ 24/40 Glass set: w/o int.: I-300 Pro, valv e unit 1 * 1 items
Catalog # BUCH11R300212N051
Supplier:  BUCHI
Frequently Bought Together