ContactUsLinkComponent
Rotavapor R-300 B-301, SJ29/32, C, P+G, I-300P, VU, 230V. . Elec. lift, B-301 bath, SJ 29/32 Glass set: Cold trap, P+G coating, int .: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300211C151
Supplier:  BUCHI
Rotavapor R-300 B-301, SJ29/32, C, P+G, I-300P, VU, 230V. . Elec. lift, B-301 bath, SJ 29/32 Glass set: Cold trap, P+G coating, int .: I-300 Pro, valve unit 1 * 1 items
Catalog # BUCH11R300211C151
Supplier:  BUCHI
Frequently Bought Together