ContactUsLinkComponent
The cost efficient Autoclave for standard sterilisation at 121°C (15 Min. holding period). Available in 12 or 18 litre volume. Printer port for CertoClav receipt printer 1 * 1 items
Catalog # 481-0825
Supplier:  CERTOCLAV
The cost efficient Autoclave for standard sterilisation at 121°C (15 Min. holding period). Available in 12 or 18 litre volume. Printer port for CertoClav receipt printer 1 * 1 items
Catalog # 481-0825
Supplier:  CERTOCLAV
Frequently Bought Together