Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
NUTRISAFE HAUTSCHUTZCREME 1L FALTFLASCHE 1 * 6 ST
Catalog # FAWE13457-007
Supplier:
FAWECO HAUTSCHUTZ
Expand image
NUTRISAFE HAUTSCHUTZCREME 1L FALTFLASCHE 1 * 6 ST
Catalog # FAWE13457-007
Supplier:
FAWECO HAUTSCHUTZ
Frequently Bought Together
Gloves, nitrile/neoprene, Microflex® 93-260
Supplier: Ansell
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
VWR® Nitrile Light Long, Nitrile Gloves
Supplier: VWR Collection
Colour Lab Tapes
Supplier: GA
VWR®, Latex Gloves
Supplier: VWR Collection
Labels, hazardous substance, GHS pictograms
Supplier: Brady
VWR® Basic Cleanroom Lab Coats, SPP, Disposable
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
High temperature anti-static cleanroom tapes
Supplier: ULTRATAPE
VWR® Nitrile Green Nitrile gloves
Supplier: VWR Collection