ContactUsLinkComponent
Maintenance-free pH sensor for harsh chemical environments, Cleaning: SIP, Autoclavation, no cable, S8 connector head, Diaphragm: 2 x Single Pore, Material: glass shaft, Electrolyte: Polisolve Plus polymeric electrolyte, Length: 425 mm, Diameter: 12 1 * 1 items
Catalog # HAMI242428-1253
Supplier:  HAMILTON BONADUZ
Maintenance-free pH sensor for harsh chemical environments, Cleaning: SIP, Autoclavation, no cable, S8 connector head, Diaphragm: 2 x Single Pore, Material: glass shaft, Electrolyte: Polisolve Plus polymeric electrolyte, Length: 425 mm, Diameter: 12 1 * 1 items
Catalog # HAMI242428-1253
Supplier:  HAMILTON BONADUZ
Frequently Bought Together