ContactUsLinkComponent
Maintenance-free pH sensor for harsh chemical environments, Cleaning: SIP, Autoclavation, no cable, S8 Connector head, Diaphragm: 2 x Single Pore, Material: glass shaft, Electrolyte: Polisolve Plus polymeric electrolyte, Length: 225 mm, Diameter: 12 1 * 1 items
Catalog # HAMI242428-1223
Supplier:  HAMILTON BONADUZ
Maintenance-free pH sensor for harsh chemical environments, Cleaning: SIP, Autoclavation, no cable, S8 Connector head, Diaphragm: 2 x Single Pore, Material: glass shaft, Electrolyte: Polisolve Plus polymeric electrolyte, Length: 225 mm, Diameter: 12 1 * 1 items
Catalog # HAMI242428-1223
Supplier:  HAMILTON BONADUZ
Frequently Bought Together