ContactUsLinkComponent
Metal gold screw lid with flip-panel Ø 43mm for twist-off glass bottle but this lid is only for Pasteurization not for Sterilisation 1 * 12 items
Catalog # SPVAR27843
Supplier:  GRAVIS
Metal gold screw lid with flip-panel Ø 43mm for twist-off glass bottle but this lid is only for Pasteurization not for Sterilisation 1 * 12 items
Catalog # SPVAR27843
Supplier:  GRAVIS
Frequently Bought Together