ContactUsLinkComponent
Model 3510 pH/temp meter with dual display, mV range and 32 reading memory, supplied without an electrode. Supplied with electrode stand and holder, ATC probe (027 500), BNC shorting plug, pH 4, 7 & 10 bu ffer sachets, UK power supply and operating i 1 * 1 items
Catalog # 662-1994
Supplier:  Cole-Parmer
Model 3510 pH/temp meter with dual display, mV range and 32 reading memory, supplied without an electrode. Supplied with electrode stand and holder, ATC probe (027 500), BNC shorting plug, pH 4, 7 & 10 bu ffer sachets, UK power supply and operating i 1 * 1 items
Catalog # 662-1994
Supplier:  Cole-Parmer
Frequently Bought Together