2-(4-Bromophenyl)-1,3-dioxolane 98+%
Glass bottle, 5 g
: 4-Bromobenzaldehyde Ethylene Acetal
- :10602-01-4
- :C₉H₉BrO₂
- :MFCD01318950
- :229.07 g/mol
- :Refrigerator
- Appearance:Colorless Liquid or low melting solid
- Infrared spectrum:Conforms
- Refractive index:1.5630 to 1.5650 (20°C, 589 nm)
- GC:≥98.0 %
- Pack type:Glass bottle
- Pk:5 g



