Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Cyclohexanone 99+% ACS

Cyclohexanone 99+% ACS

Glass bottle, 1 L
Catalog # 406090010.
Irritant
Corrosive
Flammable
   Safer Human and Environmental Health
Cyclohexanone 99+% ACS
Catalog # 406090010.
   Safer Human and Environmental Health
CAS Number:  108-94-1
Cyclohexanone 99+% ACS
Cyclohexanone 99+% ACS

Glass bottle, 1 L

CAS Number:  108-94-1
Price
2 700,00 Kč
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Boiling Pt:155 °C (760 mmHg)
  • Melting Pt:–47 °C
  • Density:0.947 g/cm³
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • UN:1915
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear viscous liquid
  • Water:
    ≤0.05 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.05 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Cat. No.:
    406090010.
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.