Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Hydroquinone 99.5%

Plastic bottle, 250 g
Catalog # ACRO219932500
Irritant
Health
Corrosive
Environment
Hydroquinone 99.5%
Catalog # ACRO219932500
CAS Number:  123-31-9
Hydroquinone 99.5%
Hydroquinone 99.5%

Plastic bottle, 250 g

CAS Number:  123-31-9
Price
1 370,00 Kč
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  1,4-Dihydroxybenzene, Quinol
  • CAS Number:123-31-9
  • Formula:C₆H₄(OH)₂
  • MDL Number:MFCD00002339
  • Molecular Weight:110.11 g/mol
  • Merck Index:13,04833
  • Boiling Pt:285…287 °C (760 mmHg)
  • Melting Pt:170…174 °C
  • Density:1,332 g/cm³ (20 °C)
  • Flash Pt:165 °C
  • Storage Temperature:Ambient
  • EINECS:204-617-8
  • UN:3077
  • ADR:9,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or crystals or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    171 to 174 °C
  • Water:
    ≤0.6 % (K.F.)
  • Sulfated ash:
    ≤0.05 %
  • HPLC:
    ≥99.4 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g

Specifications