Potassium acetate 99+% for analysis
Plastic drum, 5 kg
Dodavatel: Thermo Fisher Scientific
Potassium acetate 99+% for analysis
Katalogové číslo ACRO217100050
Dodavatel: Thermo Fisher Scientific
Číslo CAS: 127-08-2
Přejít na:
Product Details & Documents
Synonyma: Acetic acid potassium salt
- Číslo CAS:127-08-2
- Vzorec:CH₃COOK
- MDL číslo:MFCD00012458
- Molecular Weight:98.14 g/mol
- Merck Index:13,07687
- Bod tání:292 °C
- Hustota:1,57 g/cm³ (25 °C)
- Bod vzplanutí:>250 °C
- Teplota pro skladování:Ambient
- EINECS:204-822-2
Documents
ACRO21710
SDS
Specifikace
Výsledky testů specifikací
- Appearance:White Granular powder
- Infrared spectrum:Conforms
- Water:≤1.0 %
- pH:7.5 to 9.0 (5% soln. at 20°C)
- Titration with HClO4:99.0 to 101.0 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤10 ppm
Specifikace
- Pack type:Plastic drum
- Pk:5 kg
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



