Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Xylitol 99+%

Xylitol 99+%

Plastic bottle, 2,5 kg
Catalog # ACRO225980025
Xylitol 99+%
Catalog # ACRO225980025
CAS Number:  87-99-0
Xylitol 99+%
Xylitol 99+%

Plastic bottle, 2,5 kg

CAS Number:  87-99-0
Price
25 030,00 Kč
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
  • CAS Number:87-99-0
  • Formula:C₅H₁₂O₅
  • MDL Number:MFCD00064292
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,10140
  • Boiling Pt:215…217 °C
  • Melting Pt:92…96 °C
  • Density:1,52 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:201-788-0

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    92 to 96 °C
  • Water:
    ≤0.5 %
  • Sulfated ash:
    ≤0.5 %
  • GC:
    ≤2.0 % Other polyols
  • Arsenic (As):
    ≤3 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Reducing sugars:
    ≤0.2 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg

Specifications