ContactUsLinkComponent
Vacuum cleaner CWR10(4W) MRAC 240V from Tiger-Vac - ULPA Filter - Included with an efficiency of 99.999% on 0.12 micron. Tested: IEST RP-CC001.3. Filtration efficiency of 99.9995% on 0.18 micron. Tested: IEST RP-CC001.3. U15 by MPPS method as per E - 1 * 1 items
Catalog # TIGV110520B
Supplier:  TIGER-VAC
Vacuum cleaner CWR10(4W) MRAC 240V from Tiger-Vac - ULPA Filter - Included with an efficiency of 99.999% on 0.12 micron. Tested: IEST RP-CC001.3. Filtration efficiency of 99.9995% on 0.18 micron. Tested: IEST RP-CC001.3. U15 by MPPS method as per E - 1 * 1 items
Catalog # TIGV110520B
Supplier:  TIGER-VAC
Frequently Bought Together