ContactUsLinkComponent
Extraction heating unit, serial heating WHM, w. analog controller for 6 extractions for flasks flat bottom of 250/300 ml, maximum temperature 450C, external 800 x 240 x 110 mm, power 840 W, power supply: 230 V, 50/60 Hz, 1 * 1 items
Catalog # WITGDH.WHM12291
Supplier:  witeg Labortechnik
Extraction heating unit, serial heating WHM, w. analog controller for 6 extractions for flasks flat bottom of 250/300 ml, maximum temperature 450C, external 800 x 240 x 110 mm, power 840 W, power supply: 230 V, 50/60 Hz, 1 * 1 items
Catalog # WITGDH.WHM12291
Supplier:  witeg Labortechnik
Frequently Bought Together