ContactUsLinkComponent
IQ/OQ/PQ CHECK LIST WORKS TEST DATA FOR ONE FREE-SELECTABLE TEMPERATURE VALUE INCL. TEMPERATURE DISTRIBUTION SURVEY FOR 27 MEASURING POINTS TO DIN 12880:2007-5 FOR UNplus 1 * 1 items
  MEMMD00127UN110PLS
 :  MEMMERT
IQ/OQ/PQ CHECK LIST WORKS TEST DATA FOR ONE FREE-SELECTABLE TEMPERATURE VALUE INCL. TEMPERATURE DISTRIBUTION SURVEY FOR 27 MEASURING POINTS TO DIN 12880:2007-5 FOR UNplus 1 * 1 items
  MEMMD00127UN110PLS
 :  MEMMERT
Frequently Bought Together