ContactUsLinkComponent
NucleoSpin 96 Plasmid (4x 96) 4x 96 preps for the isolation of plasmid DNA-NucleoSpin Plasmid Filter Plates, NucleoSpin Plasmid Bind ing Plates, MN Wash Plates, Culture Plates, Elution Plates U-bottom, Gas-permeable Foils, Self-adhering Foils, buffer 1 * 384 items
Catalog # MANA740625.4
Supplier:  MACHEREY-NAGEL
undefined
NucleoSpin 96 Plasmid (4x 96) 4x 96 preps for the isolation of plasmid DNA-NucleoSpin Plasmid Filter Plates, NucleoSpin Plasmid Bind ing Plates, MN Wash Plates, Culture Plates, Elution Plates U-bottom, Gas-permeable Foils, Self-adhering Foils, buffer 1 * 384 items
Catalog # MANA740625.4
Supplier:  MACHEREY-NAGEL
Frequently Bought Together