ContactUsLinkComponent
Aerosol Filter Pipet Tips 10uL Low Retention. Rnase/Dnase, Endotoxin, DNA free. Tips fit most 10ul pipettors. In hinged rack. Tips are autoclavable. 96 tips per rack, 12 racks per pack. 10 packs per case. 11520 tips per case. Sterile 1 * 1 SET
Catalog # VWRI732-1486BOM
Supplier:  VWR Collection
Aerosol Filter Pipet Tips 10uL Low Retention. Rnase/Dnase, Endotoxin, DNA free. Tips fit most 10ul pipettors. In hinged rack. Tips are autoclavable. 96 tips per rack, 12 racks per pack. 10 packs per case. 11520 tips per case. Sterile 1 * 1 SET
Catalog # VWRI732-1486BOM
Supplier:  VWR Collection
Frequently Bought Together