0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Planschliffdeckel DN 100, 2xNS 29/32,3xNS 14/23 1 * 1 items
Catalog # WITG0.950.015
Supplier:
witeg Labortechnik
Expand image
Planschliffdeckel DN 100, 2xNS 29/32,3xNS 14/23 1 * 1 items
Catalog # WITG0.950.015
Supplier:
witeg Labortechnik
Frequently Bought Together
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
VWR®, Funnels with Short Stem
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
VWR®, Beakers, Tall Form
Supplier: VWR Collection
VWR®, Beakers, Low Form
Supplier: VWR Collection
VWR®, Pasteur Pipettes
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences