ContactUsLinkComponent
pH sensor with pre-pressurized electrolyte, Cleaning: SIP, CIP, Autoclavation, no cable, Connector head: VP 6, Integrated temperatur e sensor: Pt1000, Diaphragm: HP Coatramic, Material: glass shaft, Electrolyte: Phermlyte, Length: 225 mm, Diameter: 1 1 * 1 items
Catalog # HAMI243647
Supplier:  HAMILTON BONADUZ
pH sensor with pre-pressurized electrolyte, Cleaning: SIP, CIP, Autoclavation, no cable, Connector head: VP 6, Integrated temperatur e sensor: Pt1000, Diaphragm: HP Coatramic, Material: glass shaft, Electrolyte: Phermlyte, Length: 225 mm, Diameter: 1 1 * 1 items
Catalog # HAMI243647
Supplier:  HAMILTON BONADUZ
Frequently Bought Together