Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Formaldehyde 3.7-4.0% buffered to pH=7 and stabilized with methanol for clinical diagnosis 1 * 1.000 mL
Catalog # APPC252931.1211
Supplier:
PanReac AppliChem
Expand image
Formaldehyde 3.7-4.0% buffered to pH=7 and stabilized with methanol for clinical diagnosis 1 * 1.000 mL
Catalog # APPC252931.1211
Supplier:
PanReac AppliChem
Frequently Bought Together
Phosphate buffered saline, hematology reagent, for diagnostics use 1 * 10 L
Supplier: Avantor
BOND POLYMER REFINE RED DETECTION 1 * 1 KIT
Supplier: LEICA MICROSYSTEMS
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
DEHA Test Kit 1 * 1 items
Supplier: Lovibond
Nitrite VHR L, 500 ml
Supplier: Lovibond