Sodium Chloride for analysis, ACS, ISO 1 * 500 g
APPC131659.1210
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
1,10-Phenanthroline monohydrate ≥99.0% analytical reagent
: VWR Chemicals
epT.I.P.S.® Racks - Pipette Tips
: EPPENDORF
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Disposable Cuvettes
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Ammonia solution ∼32%, GPR RECTAPUR®
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals