Sodium Acetate 3-hydrate for analysis, ACS, ISO 1 * 1.000 g
Catalog # APPC131632.1211
Supplier: PanReac AppliChem
Sodium Acetate 3-hydrate for analysis, ACS, ISO 1 * 1.000 g
Catalog # APPC131632.1211
Supplier: PanReac AppliChem
Frequently Bought Together
EDTA disodium salt dihydrate BP, Ph. Eur., ACS, JP, USP
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Base materials for microbiology media
Supplier: THERMO FISHER DIAGNOSTICS