2-PROPANOL TECHNICAL GRADE 1 * 10 L
Catalog # APPC211090.0715
Supplier: PanReac AppliChem
Frequently Bought Together
Aceton ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitril, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Voda, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Supplier: Merck
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetic acid:chloroform (3:2) (v/v), pure
Supplier: PanReac AppliChem