Tin(II) Chloride 2-hydrate (Reag. Ph. Eur.) for analysis, ACS 1 * 250 g
APPC131303.1609
Tin(II) Chloride 2-hydrate (Reag. Ph. Eur.) for analysis, ACS 1 * 250 g
APPC131303.1609
Frequently Bought Together
Magnesium sulphate heptahydrate ≥98%, purified
: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
Potassium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Sodium salicylate ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Calcium chloride dihydrate 97.0-103.0% Ph. Eur.
: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals