Barium Chloride 2-hydrate (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 500 g
Catalog # APPC131182.1210
Supplier: PanReac AppliChem
Barium Chloride 2-hydrate (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 500 g
Catalog # APPC131182.1210
Supplier: PanReac AppliChem
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Test kits, chromate, Spectroquant®, Supelco®
Supplier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Potassium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium salicylate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Accessories for Plum eyewash stations
Supplier: Plum Safety