EDTA DISODIUM SALT 0.01 MOL/L (0.01M) VO
Catalog # APPC181671.1211
Supplier: PanReac AppliChem
Frequently Bought Together
Nitric acid 68%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Accessories for MA series
Supplier: Sartorius Balances
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ammonium chloride ≥99.5% ACS
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Ladies' Coats, Cotton or Polyester/Cotton
Supplier: VWR Collection
tert-Butyl methyl ether ≥99.0% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Conductivity standards
Supplier: Hach
Buffered Sodium Chloride-Peptone solution (Ph. Eur.) (Dehydrated Culture Media) for microbiology 1 * 500 g
Supplier: PanReac AppliChem