ContactUsLinkComponent
Tip filter 50µl Hamilton CO-RE style liquid level sensing sterile (96 tips/rack, 5 racks/pack, 12 packs/case) 1 * 5.760 items
Catalog # AXYGHTF-50CBKHTRS5
Supplier:  Corning
Tip filter 50µl Hamilton CO-RE style liquid level sensing sterile (96 tips/rack, 5 racks/pack, 12 packs/case) 1 * 5.760 items
Catalog # AXYGHTF-50CBKHTRS5
Supplier:  Corning
Frequently Bought Together